Learn More
Lovibond™ DPD No. 3 Rapid Tablets
Convenient blister packaging allows user to push tablet through foil directly into sample
Brand: Lovibond™ 511290BT
Additional Details : CAS : 7681-11-0 Weight : 1.00000kg
Description
Convenient blister packaging allows user to push tablet through foil directly into sample
DPD Tablet No. 1 is used to measure free chlorine; after that, DPD Tablet No. 3 is used to measure total chlorine.
- Each tablet hermetically sealed in individual aluminum foil pocket that protects it from environmental conditions for longer shelf life
- For use with Chlorine Aqua Comparator™ Test Kits
- Range: 0 to 1mg/L
Specifications
White | |
6.3 | |
Water analysis | |
0 to 1mg/L | |
DPD No. 3 Rapid Tablet |
Potassium Iodide 10 to 20%, non-hazardous components 80 to 90%, Cellulose 0.1 to 1% | |
Tablet | |
Colorimeter and spectrophotometer | |
DPD |