Learn More
Lenz Laborglasinstrumente™ Delivery Adapter, Long, Bent
For feeding distillate from condensers into receiving flasks. Lenz Laborglasinstrumente™ Delivery Adapters, Long, Bent are manufactured from durable DURAN™ glass tubing.
19.88€ - 21.78€
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
11816164
|
Lenz Laborglasinstrumente™
05263001 |
NS 14/23 |
19.88€
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11896074
|
Lenz Laborglasinstrumente™
05263002 |
NS 19/26 |
20.14€
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11876124
|
Lenz Laborglasinstrumente™
05263003 |
NS 24/29 |
21.73€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11826114
|
Lenz Laborglasinstrumente™
05263004 |
NS 29/32 |
21.78€
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
- With NS socket
- Long bent tube with angle of 105°
- 12mm diameter
Specifications
Long Bent Delivery Adapter | |
Duran Glass |