Learn More
Thermo Scientific™ LN2 Supply Tanks
Transport cryogenic industrial gases with these LN2 Supply tanks, which are designed to be both durable and user-friendly.
Brand: Thermo Scientific™ 8121
Additional Details : Weight : 196.00000kg
Description
For long-term storage of samples in cryogenic freezers, or controlling temperature during freezing procedures utilizing controlled-rate freezing equipment.- Rated at 22psi (151.7kPa)
- Evaporate 2% of volume per day
Specifications
230 L | |
66 cm | |
139.2 cm | |
LN2 Supply Tank |