missing translation for 'onlineSavingsMsg'
Learn More

Lithium bromide, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™

Catalog Number p-7111975
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7550-35-8
BrLi
86.844
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
This item is not returnable. View return policy
7550-35-8
BrLi
86.844
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]

Free-flowing, Reagent Grade, anhydrous,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Boiling Point 1,265°C
Melting Point 550°C (literature)
CAS Max % 100.0000%
Quantity 100 g
Linear Formula LiBr
Formula Weight 86.85
Percent Purity ≥99%
Packaging Poly Bottle
Density 3.46 g/cm3
Chemical Name or Material Lithium bromide, Free-flowing, Reagent Grade, anhydrous, ≥99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'