missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Lithium bromide, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™

Free-flowing, Reagent Grade, anhydrous,≥99%

Brand:  Honeywell Fluka™ 746479-6X500G

Additional Details : CAS : 7550-35-8 Weight : 3.00000kg

Packaging Poly Bottle
Quantity 6 x 500g

 View more versions of this product

Product Code. 15627020

  • 1945.40€ / Pack of 6


please call +351214253350
Add to basket

Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

7550-35-8
86.844
AMXOYNBUYSYVKV-UHFFFAOYSA-M
82050
lithium;bromide
BrLi
MFCD00011077
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
CHEBI:63042
[Li+].[Br-]

Specifications

Lithium bromide
7550-35-8
BrLi
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium;bromide
82050
86.85
3.46 g/cm3
1,265°C
6 x 500g
Free-flowing, Reagent Grade, anhydrous, ≥99%
100.0000%
LiBr
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
[Li+].[Br-]
86.844
CHEBI:63042
≥99.0%
Poly Bottle
550°C (literature)
Videos
SDS