Learn More
Abeba™ Light 1158 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1158/43
Additional Details : Weight : 0.95900kg
 View more versions of this product
Product Code. 15627137
  • 72.20€ / 1 pair
Estimated Shipment
03-08-2020
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRA
  • Leather with Air-Mesh textile inlays
  • White
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRA
Leather
Slip-resistant
1158
White
43
Polyurethane