Learn More
Buerkle™ LDPE Tall-form Bottle
Click to view available options
Capacity (Metric):
100 mL
250 mL
1000 mL
Description
- LDPE, transparent
- Leakproof
- With screw cap
- Matching dropper cap is available as accessory
Specifications
Closure Type | Screw Cap |
Height (Metric) | 198 mm |
Capacity (Metric) | 1000 mL |
Color | Transparent |
Diameter (Metric) Inner Neck | 21.6 mm |
Material | LDPE |
Thread Size (Metric) | 28 mm |
Closure Included | Yes |