Learn More
Labconco™ WaterPro™ Filters
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
106.00€ - 314.00€
Product Code | Brand | For Use With (Equipment) | Filter Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Filter Type | Price | Quantity & Availability | ||||
16297860
|
Labconco™
9007201 |
WaterPro™ PS polishing station Series 90005, 90006 and 90007 model | Activated Carbon |
184.00€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
16207870
|
Labconco™
9007301 |
WaterPro™ PS polishing station Series 90005, 90006 and 90007 model | Deionization cartridge |
274.00€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
11844750
|
Labconco™
9092700 |
WaterPro™ mobile system (Labconco Cat. Nos. 90900-00, -01, 90901-00, -01 and 90902-00, -01) | Replacement prefilter |
106.00€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
16217870
|
Labconco™
9053300 |
WaterPro™ PS/HPLC polishing station (Labconco Cat. Nos. 90006-00 to -03) and HPLC/UF Hybrid (Labconco Cat. Nos. 90007-04, -05) | Organic adsorption cartridge |
314.00€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
Description
Activated Carbon Cartridge aids in removal of contaminants during the creation of Type I ultra pure water. For use with Labconco WaterPro PS water Purification Systems.Specifications
WaterPro Filters |