missing translation for 'onlineSavingsMsg'
Learn More

L-Selenocystine, 98%, Thermo Scientific Chemicals

Catalog Number p-191786
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
29621-88-3
C6H12N2O4Se2
334.11
MFCD00800971
JULROCUWKLNBSN-IMJSIDKUSA-N
l-selenocystine, seleno-l-cystine, l-3,3'-diselenodialanine, alanine, 3,3'-diselenodi-, l, selenocystine, l, l-alanine,3,3'-diselenobis, 2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid, l-selenocystine monohydrate, selenocystine, l-isomer
207306
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]diselanyl]propanoic acid
N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O
This item is not returnable. View return policy
29621-88-3
C6H12N2O4Se2
334.11
MFCD00800971
JULROCUWKLNBSN-IMJSIDKUSA-N
l-selenocystine, seleno-l-cystine, l-3,3'-diselenodialanine, alanine, 3,3'-diselenodi-, l, selenocystine, l, l-alanine,3,3'-diselenobis, 2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid, l-selenocystine monohydrate, selenocystine, l-isomer
207306
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]diselanyl]propanoic acid
N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 212°C to 215°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble in water
Formula Weight 334.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Selenocystine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'