missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutathione, oxidized, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
L-Glutathione is used as a Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH, it is used to reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Glutathione is used as a Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH, it is used to reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress
Solubility
Soluble in water, DMF, and ethanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
L-Glutathione is used as a Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH, it is used to reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress
Solubility
Soluble in water, DMF, and ethanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Color | White |
Quantity | 1 g |
Beilstein | 1718700 |
Merck Index | 14,4475 |
Solubility Information | Soluble in water,DMF,and ethanol. |
Optical Rotation | −105° (c=2 in Water) |
Formula Weight | 612.64 |
Physical Form | Powder |
Chemical Name or Material | L-Glutathione, oxidized |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'