Learn More
L(-)-2-Amino-3-phenyl-1-propanol, 98%, Thermo Scientific Chemicals
Catalog Number p-3777300
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3182-95-4
C9H13NO
151.21
MFCD00004732
STVVMTBJNDTZBF-VIFPVBQESA-N
l-phenylalaninol, s-2-amino-3-phenyl-1-propanol, s---2-amino-3-phenyl-1-propanol, s-2-amino-3-phenylpropan-1-ol, h-phenylalaninol, h-phe-ol, 2s-2-amino-3-phenylpropan-1-ol, phenylalaninol, --l-phenylalaninol, l-phenyl alaninol
447213
(2S)-2-amino-3-phenylpropan-1-ol
N[C@H](CO)CC1=CC=CC=C1
3182-95-4
C9H13NO
151.21
MFCD00004732
STVVMTBJNDTZBF-VIFPVBQESA-N
l-phenylalaninol, s-2-amino-3-phenyl-1-propanol, s---2-amino-3-phenyl-1-propanol, s-2-amino-3-phenylpropan-1-ol, h-phenylalaninol, h-phe-ol, 2s-2-amino-3-phenylpropan-1-ol, phenylalaninol, --l-phenylalaninol, l-phenyl alaninol
447213
(2S)-2-amino-3-phenylpropan-1-ol
N[C@H](CO)CC1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 90°C to 94°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2CH(NH2)CH2OH
Quantity 1 g
Specific Rotation -23
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol,,soluble in chloroform
Formula Weight 151.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-2-Amino-3-phenyl-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-2-Amino-3-phenyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.