missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ KRT18 Recombinant Protein (P01)

335.00€ - 508.00€

Specifications

For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Molecular Weight (g/mol) 72.82
pH Range 8
Preparation Method In vitro wheat germ expression system
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16117241
View Documents Promotion Details
Abnova™
H00003875-P01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16127241
View Documents Promotion Details
Abnova™
H00003875-P01.25UG
25μg
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

Antibody Production, ELISA, Protein Array, Western Blot
72.82
In vitro wheat germ expression system
Wheat Germ (in vitro)
Recombinant
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
8
Glutathione Sepharose 4 Fast Flow
Human
Solution
Videos
SDS
Documents

Documents

Product Certifications