Learn More
DWK Life Sciences Kimble™ Kontes™ Schlenk Vacuum Tube with HI-VAC Valve
Suitable for use with K216050 or K216060 Manifolds
Brand: DWK Life Sciences 218710-0350
Additional Details : Weight : 0.80000kg
Description
- Medium wall borosilicate glass
Specifications
USP Type I, ASTM E438, Type I, Class A | |
5.1 cm | |
25 cm | |
216050 or 216060 manifolds | |
25 cm |
Borosilicate Glass | |
9.84 in. | |
Vacuum Tube | |
9 cm | |
Hi-Vac™ valve, valve plug |