Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Polyethylene Sample Tube Pressure Caps for 5 and 10mm Tubes
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporization pressures
31.90€
Specifications
Product Type | Pressure Cap |
---|---|
Material | Polyethylene |
For Use With (Application) | Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures |
Specifications
Pressure Cap | |
Polyethylene | |
Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures |