Learn More
DWK Life Sciences Kimble™ Kontes™ Clamps
For 90mm filter
Brand: DWK Life Sciences 953753-0090
Additional Details : Weight : 0.80000kg
Description
- Component part for all-glass ultra-ware microfiltration apparatus with stainless steel support
Specifications
Aluminum Clamp | |
All-glass ultra-ware microfiltration apparatus with stainless steel support | |
Aluminum | |
90 mm |