Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Brand: DWK Life Sciences 608121940
Additional Details : Weight : 0.01000kg
Description
- Manufactured from clear 33 expansion borosilicate glass
Specifications
Clear | |
Borosilicate Glass | |
Screw | |
Screw Cap | |
ASTM Type I, Class A |
19 mm | |
6 mL | |
15-425 | |
40 mm | |
15-425 |