Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials with Attached White Rubber Lined Closures
Attached closed top black phenolic closures
Brand:  DWK Life Sciences 60911D5
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11882953
  • 177.00€ / Pack of 72
Estimated Shipment
22-02-2021
Add to basket
Description
Description
  • Manufactured from amber 51 expansion borosilicate glass
  • Closures with white rubber liners
  • Vials packaged in corrugated trays with partitions
Specifications
Specifications
Amber
Borosilicate Glass
Rubber
Phenolic
57 mm
28 mm
20 mL
Screw
Screw Cap (Closed-Top)
USP Type I