Learn More
DWK Life Sciences Kimble™ Aluminum Seal Caps
With PTFE-faced rubber septa
Brand: DWK Life Sciences N7382613
Additional Details : Weight : 0.01000kg
Description
- Moderate to good resealability
- One piece
- PTFE-faced red rubber septa
- For general GC applications
- Moderate resealability
- Not recommended for multiple injections
- PTFE-faced natural rubber septa
- Good resealability
- Multiple injections
Specifications
PTFE/Rubber | |
Multiple injection | |
1000 Cs. |
Red | |
Aluminum | |
13 |