Learn More
DWK Life Sciences Kimble™ Aluminum Seal Caps
With PTFE-faced rubber septa
Brand:  DWK Life Sciences N7382613
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11853043
  • 995.00€ / Pack of 1000
Estimated Shipment
22-06-2021
Add to basket
Description
Description
  • Moderate to good resealability
  • One piece
  • PTFE-faced red rubber septa
  • For general GC applications
  • Moderate resealability
  • Not recommended for multiple injections
  • PTFE-faced natural rubber septa
  • Good resealability
  • Multiple injections
Specifications
Specifications
PTFE/Rubber
Multiple injection
1000 Cs.
Red
Aluminum
13