Learn More
DWK Life Sciences Kimble™ 51 Expansion Borosilicate Glass Melting Point Capillaries with Smaller I.D.
51 expansion borosilicate glass
Brand: DWK Life Sciences 34507-99
Additional Details : Weight : 0.01000kg
Description
- I.D.: 0.8 to 1.1mm
- 100 capillaries per vial
Specifications
0.25 mm | |
Borosilicate Glass | |
One end sealed | |
3.54 in. |
Melting Point Capillary | |
USP type I, ASTM E438 | |
0.8 to 1.1 mm | |
90 mm |