Learn More
DWK Life Sciences Kimble™ 2mL and 4mL Autosampler Vials without Closures: Crimp Openings
Constructed of borosilicate glass
Brand:  DWK Life Sciences 331232C
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11863043
  • 660.00€ / Pack of 2000
Estimated Shipment: 07-10-2024
Description
  • O.D. x Length: 12 x 32mm
  • Finish: Crimp
Specifications
Specifications
Clear
Borosilicate Glass
Chromatography applications
Regular
Crimp Cap
100/Pk.
12 mm
2.0 mL
1.25 in.
100
32 mm