Learn More
BINDER™ Keyboard Lock
For Use With: BINDER units M 53, M 115, M 240, M 400, M 720
Brand: BINDER™ 8012-0529
Additional Details : Weight : 0.10000kg
Description
- Lock and unlock the keyboard of controllers via a key switch
- To be positioned on the lateral control panel
Specifications
Lockable controller keyboard - Lockable controller keyboard with key-operated switch | |
BINDER Chambers M 53, M 115, M 240, M 400, M 720 | |
Keyboard Lock | |
Key-operated switch |