Learn More
3M™ Jupiter™ A2P Filter
Features a unique half-turn attachment, allowing an easier and safer connection. 3M™ Jupiter™ A2P Filters unit incorporates an electronic flow control within the turbo, which gives audible and visual warnings of low battery and low airflow.
Brand: 3M™ 7000006598
Additional Details : Weight : 0.80000kg
Description
Feature a unique half-turn attachment, allowing an easier and safer connection.- Unit incorporates all the latest safety features.
- An electronic flow control within the turbo, gives audible and visual warnings of low battery and low airflow.
- EN 12941 / CE marked.
- TH2 Category
Specifications
NiMH | |
For Light or Heavy Duty Use | |
A2P Filter | |
For Jupiter A2P |
EN 12941 / CE | |
Belt filters | |
Protective gas, organic vapor, inorganic acids, ammoniated, particles. | |
12/Pk. |