Learn More
3M™ Jupiter™ A2P Filter
Features a unique half-turn attachment, allowing an easier and safer connection. 3M™ Jupiter™ A2P Filters unit incorporates an electronic flow control within the turbo, which gives audible and visual warnings of low battery and low airflow.
Brand:  3M™ 7000006598
Additional Details : Weight : 0.80000kg
Product Code. 11721373
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
Feature a unique half-turn attachment, allowing an easier and safer connection.
  • Unit incorporates all the latest safety features.
  • An electronic flow control within the turbo, gives audible and visual warnings of low battery and low airflow.
  • EN 12941 / CE marked.
  • TH2 Category
Specifications
Specifications
NiMH
For Light or Heavy Duty Use
A2P Filter
For Jupiter A2P
EN 12941 / CE
Belt filters
Protective gas, organic vapor, inorganic acids, ammoniated, particles.
12/Pk.