Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap is specially designed for use with vacuum manifolds and Schlenk lines.
Brand: GPE Scientific™ JCF/1000
Additional Details : Weight : 0.35000kg
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap
Specifications
1000 mL | |
Vacuum manifolds and Schlenk lines | |
Fine thread tap | |
Sample Flask |