Learn More
Iron, Technical, Filings, Fine (Metal), Fisher Chemical™
Catalog Number p-7073094
Click to view available options
Quantity:
250 g
1 kg
3 kg
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
Color Gray
Melting Point 1535°C
Boiling Point 3000°C
Quantity 250 g
Formula Weight 55.84g/mol
Packaging Plastic powder jar
Particle Size 0.3 to 0.72mm
Vapor Pressure 1mmHg 1787
Form Solid