missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Iron(Iii) Oxide, Honeywell Fluka™

Powder, <5 μm,≥99%

Brand:  Honeywell Fluka™ 310050-500G

Additional Details : CAS : 1309-37-1 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 15623160

  • 524.73€ / 500g


Estimated Shipment date
from Supplier 21-06-2019
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1309-37-1
Fe2O3
MFCD00011008

Specifications

Iron(Iii) Oxide
Fe2O3
NONH for all modes of transport
≥99%
500g
1309-37-1
MFCD00011008
159.69g/mol
Powder
Videos
SDS