Learn More
Iron(III) oxide, 95%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1538.0°C |
Quantity | 25 kg |
Assay Percent Range | 94% min. (Iodometry) |
Linear Formula | Fe2O3 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in acids, soluble in alcohol and ether |
Formula Weight | 159.69 |
Percent Purity | 95% |
Grade | Pure |
Loss on Drying | 0.3% max. |
Specific Gravity | 5.24 |