missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) chloride tetrahydrate, Honeywell Fluka™
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 50 g |
Linear Formula | FeCl2 · 4H2O |
UN Number | UN3260 |
Formula Weight | 198.81g/mol |
Percent Purity | ≥99% (RT) |
Vapor Pressure | 10 mmHg (693°C) |
Chemical Name or Material | Iron(II) chloride tetrahydrate |
Safety and Handling
Product Identifier
- Iron(II) chloride tetrahydrate
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'