Learn More
Iron boride, 98% (metals basis), Thermo Scientific Chemicals
Description
Iron boride is used in the production of borated steels and borated stainless steels, deoxidizing agents, increase the hardness of steel, and alloying components of coatings applied to weld-facing electrodes. It is also used to improve abrasion resistance, wear resistance, and oxidation resistance. It is used in oil and gas refinery, chemical extraction, automotive, agricultural, stamping, textile extrusion and injection molding industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Formula Weight | 122.5 |
Percent Purity | 98% |
Odor | Odorless |
Chemical Name or Material | Iron boride |
RUO – Research Use Only