missing translation for 'onlineSavingsMsg'
Learn More

Iodopentafluorobenzene, 99%, stabilised over copper, Thermo Scientific Chemicals

Catalog Number p-4323614
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
827-15-6, 7440-50-8
C6F5I
293.96
OPYHNLNYCRZOGY-UHFFFAOYSA-N
iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
70008
1,2,3,4,5-pentafluoro-6-iodobenzene
C1(=C(C(=C(C(=C1F)F)I)F)F)F
This item is not returnable. View return policy
827-15-6, 7440-50-8
C6F5I
293.96
OPYHNLNYCRZOGY-UHFFFAOYSA-N
iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
70008
1,2,3,4,5-pentafluoro-6-iodobenzene
C1(=C(C(=C(C(=C1F)F)I)F)F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 2.2040g/mL
Boiling Point 162.0°C to 165.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.496
Quantity 1 g
Specific Gravity 2.204
Formula Weight 293.96
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Iodopentafluorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • Iodopentafluorobenzene, stabilised over copper
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'