missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodobenzene, 98%, ACROS Organics™
Brand: Acros Organics 122330025
Additional Details : Weight : 4.60000kg
Packaging | Glass bottle |
---|---|
Quantity | 2.5L |
Chemical Identifiers
591-50-4 | |
204.01 | |
benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene | |
iodobenzene |
C6H5I | |
SNHMUERNLJLMHN-UHFFFAOYSA-N | |
11575 | |
C1=CC=C(C=C1)I |
Specifications
Iodobenzene | |
74°C | |
Glass bottle | |
1.82 | |
Yellow | |
2.5L | |
97.5% min. (GC) | |
C6H5I | |
01,505 | |
benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene | |
SNHMUERNLJLMHN-UHFFFAOYSA-N | |
iodobenzene | |
11575 | |
Liquid |
1.8200g/mL | |
Authentic | |
1.6180 to 1.6200 | |
188.0°C | |
-29.0°C | |
591-50-4 | |
C6H5I | |
,215 | |
15,5072 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether | |
C1=CC=C(C=C1)I | |
204.01 | |
204.01 | |
98% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Iodobenzene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.