missing translation for 'onlineSavingsMsg'
Learn More

Iodine Solution, 0.05M, Chem-Lab

Catalog Number p-8014300
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
7553-56-2, 7732-18-5
I2
253.81
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
807
CHEBI:17606
II
This item is not returnable. View return policy
7553-56-2, 7732-18-5
I2
253.81
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
807
CHEBI:17606
II

Used in laboratories for accurate volumetric analysis and titrations. Iodine Solution, 0.05M, Chem-Lab is a standardized solution with 12.69g I2 per 1L water. It is also used topically as a disinfectant and antibacterial agent. (Calibrate before use.)

Iodine is a nonmetallic element of the halogen group. It is a nutritionally essential element, especially important in thyroid hormone synthesis. In solution, it has anti-infective properties (when used topically).
pH 5.8–6.2
Quantity 500 mL
IUPAC Name diiodine
Formula Weight 253.81g/mol
Concentration or Composition (by Analyte or Components) 0.05mol/L
Density 1.02g/mL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'