missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Puriss., ≥99.5% (RT), particles (round), Honeywell Fluka™

Catalog Number p-7111954
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
7553-56-2
I2
253.81
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
807
CHEBI:17606
II
This item is not returnable. View return policy
7553-56-2
I2
253.81
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
807
CHEBI:17606
II

Puriss.,≥99.5% (RT), particles (round)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 113°C
Color Black
Physical Form Solid
Quantity 100 g
UN Number UN3495
Beilstein 3587194
IUPAC Name diiodine
Formula Weight 253.81g/mol
Percent Purity ≥99.5% (RT)
Packaging Glass bottle
Vapor Pressure 1mmHg (38.7°C), 0.31mmHg (25°C)
Density 4.930g/mL
Chemical Name or Material Iodine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'