Learn More
ACROBiosystems Influenza A [A/Shanghai/2/2013(H7N9)] Hemagglutinin (HA) Protein, His Tag
POA
Specifications
Accession Number | AHF20558.1 |
---|---|
Molecular Weight (g/mol) | 56.7 kDa |
Source | Influenza A [A/Shanghai/2/2013(H7N9)] HA, His Tag (HA9-V5227) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Gly 338, Leu 341 - Asp 523 (Accession # R4NN21-1). |
Immunogen | Asp 19 - Gly 338, Leu 341 - Asp 523 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15836917
|
ACROBiosystems
HA9-V5227-1MG |
1 mg |
POA
1mg |
N/A | |||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15826917
|
ACROBiosystems
HA9-V5227-100UG |
100 μg |
POA
100µg |
N/A | |||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
Influenza, commonly known as “the flu”, is an infectious disease of birds and mammals caused by RNA viruses of the family Orthomyxoviridae, the influenza viruses. The virus is divided into three main types (Influenzavirus A, Influenzavirus B, and Influenzavirus C), which are distinguished by differences in two major internal proteins (hemagglutinin (HA) and neuraminidase (NA), which are the most important targets for the immune system. The type A viruses are the most virulent human pathogens among the three influenza types and cause the most severe disease. The serotypes that have been confirmed in humans, ordered by the number of known human pandemic deaths, are: H1N1, H2N2, H3N2, H5N1, H7N7, H1N2, H9N2, H7N2, H7N3, H10N7, H7N9. H7N9 is a serotype of the species Influenzavirus A (avian influenza virus or bird flu virus). H7 normally circulates amongst avian populations with some variants known to occasionally infect humans. An H7N9 virus was first reported to have infected humans in 2013 in China.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 56.7 kDa. The protein migrates as 63-75 kDa under reducing (R) condition (SDS-PAGE).Specifications
AHF20558.1 | |
Influenza A [A/Shanghai/2/2013(H7N9)] HA, His Tag (HA9-V5227) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Gly 338, Leu 341 - Asp 523 (Accession # R4NN21-1). | |
-20°C | |
Hemagglutinin (HA) | |
Influenza | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
56.7 kDa | |
Asp 19 - Gly 338, Leu 341 - Asp 523 | |
HA,Hemagglutinin | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |