Learn More
ACROBiosystems Influenza A [A/Guinea fowl/Hong Kong/WF10/99 (H9N2)] Hemagglutinin 1 (HA1) Protein, His Tag
Brand: ACROBiosystems HA2-V52H8-1MG
Additional Details : Weight : 0.00001kg
Description
Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Hemagglutinin also plays a major role in the determination of host range restriction and virulence. As a class I viral fusion protein, hemagglutinin is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.5 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
Q80KD9 | |
1 mg | |
Asp 19 - Arg 338 | |
HA1,Hemagglutinin 1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
37.5 kDa | |
Influenza A [A/Guinea fowl/Hong Kong/WF10/99 (H9N2)] HA1, His Tag (HA2-V52H8) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Arg 338 (Accession # Q80KD9). | |
-20°C | |
Hemagglutinin 1 (HA1) | |
Influenza | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |