missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imidazole, >98.5% (Dry Basis), Ultrapure, Thermo Scientific™

Brand:  Thermo Scientific™ J17525.A1

Additional Details : CAS : 288-32-4 Weight : 1.00000kg Transport : UN number : 3263 Chem class : 8 Pack group : III

Packaging Plastic bottle
Quantity 1kg

 View more versions of this product

Product Code. 15815308

  • 168.84€ / 1kg


In stock, ready to ship: 3
Add to basket

Description

Description

Specifications

Chemical Identifiers

288-32-4
68.079
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
1H-imidazole
C3H4N2
MFCD00005183
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
CHEBI:16069
C1=CN=CN1

Specifications

Imidazole
145°C (293°F)
1.03
88°C to 92°C
288-32-4
C3H4N2
UN3263
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
C1=CN=CN1
68.079
CHEBI:16069
Flakes/Powder
Ultrapure
1.03
Plastic bottle
256°C
1kg
Dry basis
MFCD00005183
103853
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
795
68.08
≥98.5%
Videos
SDS
missing translation for 'documents'
Product Certifications