missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine, 99%, Thermo Scientific Chemicals

Catalog Number p-4229252
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
274-76-0
C7H6N2
118.14
MFCD00005553
UTCSSFWDNNEEBH-UHFFFAOYSA-N
imidazo 1,2-a pyridine, unii-g18zbv2hxa, 1h-imidazo 1,2-a pyridine, g18zbv2hxa, pyrimidazole, 1-azaindolizine, 1,3a-diazaindene, pubchem17181, imidazo l,2-a pyridine
78960
imidazo[1,2-a]pyridine
C1=CN2C=CC=CC2=N1
This item is not returnable. View return policy
274-76-0
C7H6N2
118.14
MFCD00005553
UTCSSFWDNNEEBH-UHFFFAOYSA-N
imidazo 1,2-a pyridine, unii-g18zbv2hxa, 1h-imidazo 1,2-a pyridine, g18zbv2hxa, pyrimidazole, 1-azaindolizine, 1,3a-diazaindene, pubchem17181, imidazo l,2-a pyridine
78960
imidazo[1,2-a]pyridine
C1=CN2C=CC=CC2=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Boiling Point 97°C to 98°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 118.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Imidazo[1, 2-a]pyridine
Show More Show Less
compliance-icons
Product Identifier
  • Imidazo¢1,2-a!pyridine
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'