missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL7 (Human) Recombinant Protein
Human IL7 full-length ORF ( AAH47698, 26 a.a. - 177 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH47698 |
---|---|
Gene ID (Entrez) | 3574 |
Name | interleukin 7 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16156791
|
Abnova™
H00003574-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16166791
|
Abnova™
H00003574-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
- Sequence: DCDIEGKDGKQYESVLVVSIDQLLDSMKEIGSNCLNNEFNFFKRHICDANKEGMFLFRAARKLRQFLKMNSTGDFDLHLLKVSEGTTILLNCTGQVKGRKPAALGEAQPTKSLEENKSLKEQKKLNDLCFLKRLLQEIKTCWNKILMGTKEH
Specifications
AAH47698 | |
interleukin 7 | |
125% SDS-PAGE Stained with Coomassie Blue | |
IL-7 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
3574 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL7 | |
GST |