missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-5 Rat anti-Mouse/Human, APC, Clone: TRFK5, BD
Rat Monoclonal Antibody
253.00€ - 510.00€
Specifications
Antigen | IL-5 |
---|---|
Clone | TRFK5 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15811599
|
BD Biosciences
562048 |
25 μg |
253.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15817668
|
BD Biosciences
554396 |
0.1 mg |
510.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
The TRFK5 antibody reacts with mouse interleukin-5 (IL-5) and cross-reacts with human IL-5. The TRFK5 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.
Specifications
IL-5 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Human, Mouse | |
Mouse Semi-Purified T-Cell Clone Supernatant | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
TRFK5 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.