Learn More
IL-2 Rat anti-Mouse, APC, Clone: JES6-5H4, BD
Rat Monoclonal Antibody
176.00€ - 464.00€
Specifications
Antigen | IL-2 |
---|---|
Clone | JES6-5H4 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15841589
|
BD Biosciences
562041 |
25 μg |
176.00€
Each |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
15807698
|
BD Biosciences
554429 |
0.1 mg |
464.00€
Each |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
Description
The JES6-5H4 monoclonal antibody specifically binds to mouse interleukin-2 (IL-2). IL-2 is a multifunctional cytokine that plays pivotal roles in immunity and tolerance. It is produced by activated T cells. IL-2 effects the activation, growth, proliferation and/or differentiation of various cell types including T and B lymphocytes and their precursors, LAK cells, NK cells, and monocytes/macrophages. IL-2 mediates its biological activities by binding to IL-2 receptor compleses. The intermediate affinity IL-2R is comprised of IL-2R β (CD122) and common gamma chain ( γc; CD132) subunits whereas the high-affinity IL-2R is comprised of IL-2R α (CD25), IL-2R β and γc subunits. The JES6-5H4 monoclonal antibody binds to IL-2 and neutralizes its biological activity.
Specifications
IL-2 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
IgG2b | |
Affinity Purified |
JES6-5H4 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.