Learn More
IgG (H+L) Goat anti-Mouse, Rhodamine, Polyclonal, Proteintech
Goat Polyclonal Antibody
Brand:  Proteintech SA00007-1-200UL
Product Code. 17234916
  • 50.00€ / 200µL
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human, Mouse, Rat, Pig, Rabbit samples. It is validated for Immunofluorescence, Flow Cytometry.
Specifications
Specifications
IgG (H+L)
Polyclonal
Rhodamine
200 μL
Human, Mouse, Rat, Pig, Rabbit
IgG
Immunofluorescence, Flow Cytometry
0.6 mg/mL
Goat
Secondary