missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Lambda Human, Biotin, Isotype Control, Novus Biologicals™
Human Polyclonal Isotype Control
Brand: Novus Biologicals NBP3-06874B
This item is not returnable.
View return policy
Description
IgA Lambda Polyclonal antibody specifically detects IgA Lambda in Human samples. It is validated for ELISA, ControlSpecifications
IgA λ | |
Polyclonal | |
PBS | |
Human myeloma serum | |
RUO | |
Store at 4°C in the dark. | |
IgA λ |
ELISA | |
Biotin | |
Human | |
0.1 mL | |
Human | |
Isotype Controls |