Learn More
Thermo Scientific™ BioProcess Containers for HyPerforma™ Single-Use Mixer
Support impeller-based mixing in bioproduction using these bioprocess containers.
1234.61€ - 1851.61€
Products 6
Product Code Brand Description Capacity (Metric) Price Quantity & Availability  
Product Code Brand Description Capacity (Metric) Price Quantity & Availability  
10583934 Thermo Scientific™
SH30753.01
200L S.U.M. BPC, liquid-to-liquid 200L 1234.61€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-07-2019
Add to basket
10608174 Thermo Scientific™
SH30753.02
200L S.U.M. BPC, liquid-to-liquid w/probe ports 200L 1333.96€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-07-2019
Add to basket
10593934 Thermo Scientific™
SH30754.01
500L S.U.M. BPC, liquid-to-liquid 500L 1497.67€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-07-2019
Add to basket
10656444 Thermo Scientific™
SH30754.02
500L S.U.M. BPC, liquid-to-liquid w/probe ports 500L 1631.90€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-07-2019
Add to basket
10268073 Thermo Scientific™
SH30755.01
1000L S.U.M. BPC, liquid-to-liquid 1000L 1751.95€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-07-2019
Add to basket
10791274 Thermo Scientific™
SH30755.02
1000L S.U.M. BPC, liquid-to-liquid w/probe ports 1000L 1851.61€
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-07-2019
Add to basket
Description
Description
Single Use Mixer (S.U.M.) BPC options include:
  • Open top liner for media/buffer hydration
  • Closed BPC with powder port for contained media/buffer hydration
  • Closed BPC with powder port and monitoring probe capability
  • Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
  • Closed BPC for liquid-liquid mixing with monitoring probe capability