missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZBTB7B Partial ORF (NP_056956.1, 433 a.a. - 537 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_056956.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51043 |
Molecular Weight (g/mol) | 37.29kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16136546
|
Abnova™
H00051043-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16126546
|
Abnova™
H00051043-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
ZFP67 is an early growth response gene that encodes a zinc finger-containing transcription factor that binds to the promoter regions of type I collagen genes (e.g., COL1A1; MIM 120150) and has a role in development.[supplied by OMIM]
Sequence: HLCHRAFAKEDHLQRHLKGQNCLEVRTRRRRKDDAPPHYPPPSTAAAFPAGLDLSNGHLDTFRLSLARFWEQSAPTWAPVSTPGPPDDDEEEGAPTTPQAEGAMESpecifications
NP_056956.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686G01254/THPOK/ZBTB15/ZFP67/ZNF857B/c-Krox/hcKrox | |
ZBTB7B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51043 | |
ZBTB7B (Human) Recombinant Protein (Q01) | |
HLCHRAFAKEDHLQRHLKGQNCLEVRTRRRRKDDAPPHYPPPSTAAAFPAGLDLSNGHLDTFRLSLARFWEQSAPTWAPVSTPGPPDDDEEEGAPTTPQAEGAME | |
RUO | |
ZBTB7B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |