missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human YME1L1 Full-length ORF (NP_647474.1, 1 a.a. - 740 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010730-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125622

  • 359.00€ / 10µg

Estimated Shipment: 07-10-2022
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MFSLSSTVQPQVTVPLSHLINAFHTPKNTSVSLSGVSVSQNQHRDVVPEHEAPSSECMFSDFLTKLNIVSIGKGKIFEGYRSMFMEPAKRMKKSLDTTDNWHIRPEPFSLSIPPSLNLRDLGLSELKIGQIDQLVENLLPGFCKGKNISSHWHTSHVSAQSFFENKYGNLDIFSTLRSSCLYRHHSRALQSICSDLQYWPVFIQSRGFKTLKSRTRRLQSTSERLAETQNIAPSFVKGFLLRDRGSDVESLDKLMKTKNIPEAHQDAFKTGFAEGFLKAQALTQKTNDSLRRTRLILFVLLLFGIYGLLKNPFLSVRFRTTTGLDSAVDPVQMKNVTFEHVKGVEEAKQELQEVVEFLKNPQKFTILGGKLPKGILLVGPPGTGKTLLARAVAGEADVPFYYASGSEFDEMFVGVGASRIRNLFREAKANAPCVIFIDELDSVGGKRIESPMHPYSRQTINQLLAEMDGFKPNEGVIIIGATNFPEALDNALIRPGRFDMQVTVPRPDVKGRTEILKWYLNKIKFDQSVDPEIIARGTVGFSGAELENLVNQAALKAAVDGKEMVTMKELEFSKDKILMGPERRSVEIDNKNKTITAYHESGHAIIAYYTKDAMPINKATIMPRGPTLGHVSLLPENDRWNETRAQLLAQMDVSMGGRVAEELIFGTDHITTGASSDFDNATKIAKRMVTKFGMSEKLGVMTYSDTGKLSPETQSAIEQEIRILLRDSYERAKHILKTHS
Specifications

Specifications

NP_647474.1
Liquid
10730
YME1L1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MFSLSSTVQPQVTVPLSHLINAFHTPKNTSVSLSGVSVSQNQHRDVVPEHEAPSSECMFSDFLTKLNIVSIGKGKIFEGYRSMFMEPAKRMKKSLDTTDNWHIRPEPFSLSIPPSLNLRDLGLSELKIGQIDQLVENLLPGFCKGKNISSHWHTSHVSAQSFFENKYGNLDIFSTLRSSCLYRHHSRALQSICSDLQYWPVFIQSRGFKTLKSRTRRLQSTSERLAETQNIAPSFVKGFLLRDRGSDVESLDKLMKTKNIPEAHQDAFKTGFAEGFLKAQALTQKTNDSLRRTRLILFVLLLFGIYGLLKNPFLSVRFRTTTGLDSAVDPVQMKNVTFEHVKGVEEAKQELQEVVEFLKNPQKFTILGGKLPKGILLVGPPGTGKTLLARAVAGEADVPFYYASGSEFDEMFVGVGASRIRNLFREAKANAPCVIFIDELDSVGGKRIESPMHPYSRQTINQLLAEMDGFKPNEGVIIIGATNFPEALDNALIRPGRFDMQVTVPRPDVKGRTEILKWYLNKIKFDQSVDPEIIARGTVGFSGAELENLVNQAALKAAVDGKEMVTMKELEFSKDKILMGPERRSVEIDNKNKTITAYHESGHAIIAYYTKDAMPINKATIMPRGPTLGHVSLLPENDRWNETRAQLLAQMDVSMGGRVAEELIFGTDHITTGASSDFDNATKIAKRMVTKFGMSEKLGVMTYSDTGKLSPETQSAIEQEIRILLRDSYERAKHILKTHS
RUO
YME1L1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
109.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FTSH/MEG4/PAMP/YME1L
YME1L1
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers