missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human YAF2 Full-length ORF (NP_005739.2, 1 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005739.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10138 |
Molecular Weight (g/mol) | 46.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124552
|
Abnova™
H00010138-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16134552
|
Abnova™
H00010138-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene interacts with YY1, a zinc finger protein involved in negative regulation of muscle-restricted genes. This gene product itself contains a single N-terminal C2-X10-C2 zinc finger, and in contrast to YY1, is up-regulated during myogenic differentiation. It also facilitates proteolytic cleavage of YY1 by the calcium- activated protease, m-calpain, suggesting a mechanism by which this protein antagonizes the negative effect of YY1. [provided by RefSeq]
Sequence: MGDKKSPTRPKRQPKPSSDEGYWDCSVCTFRNSAEAFKCMMCDVRKGTSTRKPRPVSQLVAQQVTQQFVPPTQSKKEKKDKVEKEKSEKETTSKKNSHKKTRPRLKNVDRSSAQHLEVTVGDLTVIITDFKEKTKSPPASSAASADQHSQSGSSSDNTERGMSRSSSPRGEASSLNGESHSpecifications
NP_005739.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGDKKSPTRPKRQPKPSSDEGYWDCSVCTFRNSAEAFKCMMCDVRKGTSTRKPRPVSQLVAQQVTQQFVPPTQSKKEKKDKVEKEKSEKETTSKKNSHKKTRPRLKNVDRSSAQHLEVTVGDLTVIITDFKEKTKSPPASSAASADQHSQSGSSSDNTERGMSRSSSPRGEASSLNGESH | |
RUO | |
YAF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10138 | |
YAF2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC41856 | |
YAF2 | |
Recombinant | |
wheat germ expression system |