missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WRB Full-length ORF (NP_004618.2, 1 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004618.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7485 |
Molecular Weight (g/mol) | 46.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16104098
|
Abnova™
H00007485-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16194088
|
Abnova™
H00007485-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a basic nuclear protein of unknown function. The gene is widely expressed in adult and fetal tissues. Since the region proposed to contain the gene(s) for congenital heart disease (CHD) in Down syndrome (DS) patients has been restricted to 21q22.2-22.3, this gene, which maps to 21q22.3, has a potential role in the pathogenesis of Down syndrome congenital heart disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSSAAADHWAWLLVLSFVFGCNVLRILLPSFSSFMSRVLQKDAEQESQMRAEIQDMKQELSTVNMMDEFARYARLERKINKMTDKLKTHVKARTAQLAKIKWVISVAFYVLQAALMISLIWKYYSVPVAVVPSKWITPLDRLVAFPTRVAGGVGITCWILVCNKVVAIVLHPFSSpecifications
NP_004618.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSSAAADHWAWLLVLSFVFGCNVLRILLPSFSSFMSRVLQKDAEQESQMRAEIQDMKQELSTVNMMDEFARYARLERKINKMTDKLKTHVKARTAQLAKIKWVISVAFYVLQAALMISLIWKYYSVPVAVVPSKWITPLDRLVAFPTRVAGGVGITCWILVCNKVVAIVLHPFS | |
RUO | |
WRB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7485 | |
WRB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHD5 | |
WRB | |
Recombinant | |
wheat germ expression system |