missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WDR62 Full-length ORF (AAH17261.1, 1 a.a. - 630 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH17261.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 284403 |
Molecular Weight (g/mol) | 93.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16169953
|
Abnova™
H00284403-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16159953
|
Abnova™
H00284403-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Sequence: MKPESLENSILDSLEPQSLASLLSESESPQEAGRGHPSFLPQQKESSEASELILYSLEAEVTVTGTDSQYCRKEVEAGPGDQQGDSYLRVSSDSPKDQSPPEDSGESEADLECSFAAIHSPAPPPDPAPRFATSLPHFPGCAGPTEDELSLPEGPSVPSSSLPQTPEQEKFLRHHFETLTESPCRALGDVEASEAEDHFFNPRLSISTQFLSSLQKASRFTHTFPPRATQCLVKSPEVKLMDRGGSQPRAGTGYASPDRTHVLAAGKAEETLEAWRPPPPCLTSLASCVPASSVLPTDRNLPTPTSAPTPGLAQGVHAPSTCSYMEATASSRARISRSISLGDSEGPIVATLAQPLRRPSSVGELASLGQELQAITTATTPSLDSEGQEPALRSWGNHEARANLRLTLSSACDGLLLPPVDTQPGVTVPAVSFPAPSPVEESALRLHGSAFRPSLPAPESPGLPAHPSNPQLPEARPGIPGSTASLLEPTSGALGLFQGSPARWSEPWVPVEALPPSPLELSRVGNILHRLQTTFQEALDLYRVLVSSGQVDTGQQQARTELVSTFLWIHSQLEAECLVGTSVAPAQALPSPGPPSPPTLYPLASPDLQALLEHYSELLVQAVRRKARGHSpecifications
AAH17261.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
93.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKPESLENSILDSLEPQSLASLLSESESPQEAGRGHPSFLPQQKESSEASELILYSLEAEVTVTGTDSQYCRKEVEAGPGDQQGDSYLRVSSDSPKDQSPPEDSGESEADLECSFAAIHSPAPPPDPAPRFATSLPHFPGCAGPTEDELSLPEGPSVPSSSLPQTPEQEKFLRHHFETLTESPCRALGDVEASEAEDHFFNPRLSISTQFLSSLQKASRFTHTFPPRATQCLVKSPEVKLMDRGGSQPRAGTGYASPDRTHVLAAGKAEETLEAWRPPPPCLTSLASCVPASSVLPTDRNLPTPTSAPTPGLAQGVHAPSTCSYMEATASSRARISRSISLGDSEGPIVATLAQPLRRPSSVGELASLGQELQAITTATTPSLDSEGQEPALRSWGNHEARANLRLTLSSACDGLLLPPVDTQPGVTVPAVSFPAPSPVEESALRLHGSAFRPSLPAPESPGLPAHPSNPQLPEARPGIPGSTASLLEPTSGALGLFQGSPARWSEPWVPVEALPPSPLELSRVGNILHRLQTTFQEALDLYRVLVSSGQVDTGQQQARTELVSTFLWIHSQLEAECLVGTSVAPAQALPSPGPPSPPTLYPLASPDLQALLEHYSELLVQAVRRKARGH | |
RUO | |
WDR62 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
284403 | |
WDR62 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C19orf14/DKFZp434J046/DKFZp686G1024/FLJ33298/MGC166976 | |
WDR62 | |
Recombinant | |
wheat germ expression system |