missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WDR5B Full-length ORF (NP_061942.2, 1 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_061942.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54554
Molecular Weight (g/mol) 62.7kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16160643
View Documents Promotion Details
Abnova™
H00054554-P01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16150643
View Documents Promotion Details
Abnova™
H00054554-P01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

This intronless gene encodes a protein containing several WD40 repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, including a trp-asp at the C-terminal end. The encoded protein may mediate protein-protein interactions. [provided by RefSeq]

Sequence: MATKESRDAKAQLALSSSANQSKEVPENPNYALKCTLVGHTEAVSSVKFSPNGEWLASSSADRLIIIWGAYDGKYEKTLYGHNLEISDVAWSSDSSRLVSASDDKTLKLWDVRSGKCLKTLKGHSNYVFCCNFNPPSNLIISGSFDETVKIWEVKTGKCLKTLSAHSDPVSAVHFNCSGSLIVSGSYDGLCRIWDAASGQCLKTLVDDDNPPVSFVKFSPNGKYILTATLDNTLKLWDYSRGRCLKTYTGHKNEKYCIFANFSVTGGKWIVSGSEDNLVYIWNLQTKEIVQKLQGHTDVVISAACHPTENLIASAALENDKTIKLWMSNH
Specifications

Specifications

NP_061942.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.7kDa
Glutathione Sepharose 4 Fast Flow
MATKESRDAKAQLALSSSANQSKEVPENPNYALKCTLVGHTEAVSSVKFSPNGEWLASSSADRLIIIWGAYDGKYEKTLYGHNLEISDVAWSSDSSRLVSASDDKTLKLWDVRSGKCLKTLKGHSNYVFCCNFNPPSNLIISGSFDETVKIWEVKTGKCLKTLSAHSDPVSAVHFNCSGSLIVSGSYDGLCRIWDAASGQCLKTLVDDDNPPVSFVKFSPNGKYILTATLDNTLKLWDYSRGRCLKTYTGHKNEKYCIFANFSVTGGKWIVSGSEDNLVYIWNLQTKEIVQKLQGHTDVVISAACHPTENLIASAALENDKTIKLWMSNH
RUO
WDR5B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
54554
WDR5B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11287/MGC49879
WDR5B
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications