missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human VSIG2 Protein, His Tag

Brand:  ACROBiosystems VS2-H5225-200UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15807947

  • 282.47€ / 200µg


Estimated Shipment: 19-10-2021
Add to basket

Description

Description

V-set and immunoglobulin domain-containing protein 2 (VSIG2) is also known as Cortical thymocyte-like protein, CTH, CTXL, is a single-pass type I membrane protein which contains Ig-like C2-type (immunoglobulin-like) domain and one Ig-like V-type (immunoglobulin-like) domain. V-set domains are Ig-like domains resembling the antibody variable domain. VSIG2 is highly expressed in stomach, colon, prostate, trachea and thyroid glands and weakly in bladder and lung. V-set domains are found in diverse protein families, including immunoglobulin light and heavy chains.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

AAH07313
HEK293
Human
24.4 kDa
200 µg
Val 24 - Ala 243
VSIG2,CT-like protein,CTH,CTXL
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>95% as determined by SDS-PAGE
Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Ala 243 (Accession # AAH07313).
-20°C
VSIG2
His Tag
Videos
SDS
Documents
Certificates