Learn More
Abnova™ Human VPS41 Full-length ORF (P49754, 1 a.a. - 854 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | P49754 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27072 |
Molecular Weight (g/mol) | 124.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128522
|
Abnova™
H00027072-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16118522
|
Abnova™
H00027072-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human ortholog of yeast Vps41 protein which is also conserved in Drosophila, tomato, and Arabidopsis. Expression studies in yeast and human indicate that this protein may be involved in the formation and fusion of transport vesicles from the Golgi. Several transcript variants encoding different isoforms have been described for this gene, however, the full-length nature of not all is known. [provided by RefSeq]
Sequence: MAEAVEQETGSLEESTDESEEEESEEEPKLKYERLSNGVTEILQKDAASCMTVHDKFLALGTHYGKVYLLDVQGNITQKFDVSPVKINQISLDESGEHMGVCSEDGKVQVFGLYSGEEFHETFDCPIKIIAVHPHFVRSSCKQFVTGGKKLLLFERSWMNRWKSAVLHEGEGNIRSVKWRGHLIAWANNMGVKIFDIISKQRITNVPRDDISLRPDMYPCSLCWKDNVTLIIGWGTSVKVCSVKERHASEMRDLPSRYVEIVSQFETEFYISGLAPLCDQLVVLSYVKEISEKTEREYCARPRLDIIQPLSETCEEISSDALTVRGFQENECRDYHLEYSEGESLFYIVSPRDVVVAKERDQDDHIDWLLEKKKYEEALMAAEISQKNIKRHKILDIGLAYINHLVERGDYDIAARKCQKILGKNAALWEYEVYKFKEIGQLKAISPYLPRGDPVLKPLIYEMILHEFLESDYEGFATLIREWPGDLYNNSVIVQAVRDHLKKDSQNKTLLKTLAELYTYDKNYGNALEIYLTLRHKDVFQLIHKHNLFSSIKDKIVLLMDFDSEKAVDMLLDNEDKISIKKVVEELEDRPELQHVYLHKLFKRDHHKGQRYHEKQISLYAEYDRPNLLPFLRDSTHCPLEKALEICQQRNFVEETVYLLSRMGNSRSALKMIMEELHDVDKAIEFAKEQDDGELWEDLILYSIDKPPFITGLLNNIGTHVDPILLIHRIKEGMEIPNLRDSLVKILQDYNLQILLREGCKKILVADSLSLLKKMHRTQMKGVLVDEENICESCLSPILPSDAAKPFSVVVFHCRHMFHKECLPMPSMNSAAQFCNICSAKNRGPGSAILEMKKSpecifications
P49754 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
124.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAEAVEQETGSLEESTDESEEEESEEEPKLKYERLSNGVTEILQKDAASCMTVHDKFLALGTHYGKVYLLDVQGNITQKFDVSPVKINQISLDESGEHMGVCSEDGKVQVFGLYSGEEFHETFDCPIKIIAVHPHFVRSSCKQFVTGGKKLLLFERSWMNRWKSAVLHEGEGNIRSVKWRGHLIAWANNMGVKIFDIISKQRITNVPRDDISLRPDMYPCSLCWKDNVTLIIGWGTSVKVCSVKERHASEMRDLPSRYVEIVSQFETEFYISGLAPLCDQLVVLSYVKEISEKTEREYCARPRLDIIQPLSETCEEISSDALTVRGFQENECRDYHLEYSEGESLFYIVSPRDVVVAKERDQDDHIDWLLEKKKYEEALMAAEISQKNIKRHKILDIGLAYINHLVERGDYDIAARKCQKILGKNAALWEYEVYKFKEIGQLKAISPYLPRGDPVLKPLIYEMILHEFLESDYEGFATLIREWPGDLYNNSVIVQAVRDHLKKDSQNKTLLKTLAELYTYDKNYGNALEIYLTLRHKDVFQLIHKHNLFSSIKDKIVLLMDFDSEKAVDMLLDNEDKISIKKVVEELEDRPELQHVYLHKLFKRDHHKGQRYHEKQISLYAEYDRPNLLPFLRDSTHCPLEKALEICQQRNFVEETVYLLSRMGNSRSALKMIMEELHDVDKAIEFAKEQDDGELWEDLILYSIDKPPFITGLLNNIGTHVDPILLIHRIKEGMEIPNLRDSLVKILQDYNLQILLREGCKKILVADSLSLLKKMHRTQMKGVLVDEENICESCLSPILPSDAAKPFSVVVFHCRHMFHKECLPMPSMNSAAQFCNICSAKNRGPGSAILEMKK | |
RUO | |
VPS41 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
27072 | |
VPS41 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HVPS41/HVSP41/hVps41p | |
VPS41 | |
Recombinant | |
wheat germ expression system |